Learn More
Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
Product Code. 11544526 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500mL
100 mL
20 x 100mL
500mL
Unit Size:
100mL
5000mL
500mL
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11544526 500mL 500mL
11534526 100 mL 100mL
11564526 20 x 100mL Pack of 20
11554526 10 x 500mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11544526 Supplier Gibco™ Supplier No. 72400021
  • ONLINE EXCLUSIVE PRICE
    367.00 NOK
    352.65 NOK / 500mL
    Save 14.35 NOK
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications.
Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. Gibco™ RPMI withGlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).
cGMP Manufacturing and Quality System
Gibco™ RPMI 1640 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ RPMI 1640 product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 X
Form Liquid
Product Type Roswell Park Memorial Institute (RPMI) 1640 Medium
With Additives GlutaMAX, Phenol Red, HEPES
Without Additives No Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Quantity 500mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7.0 to 7.4
Content And Storage 2 to 8°C
Description Roswell Park Memorial Institute (RPMI) 1640 Medium, 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glutamax™ I, Phenol Red, HEPES, 12 Months Shelf Life, 270 to 310mOsm/kg Osmolality, For Mammalian Cell Culture