Learn More
MP Biomedicals™ RPMI 1640 Medium
Product Code. p-4474858 Shop All MP Biomedicals Products
Click to view available options
pH:
6.8 to 7.4
6.9 to 7.2
6.9 to 7.4
Content And Storage:
0° to 5°C
15° to 30°C
4°C
Unit Size:
100mL
500mL
Product Code. 11400385
Brand: MP Biomedicals™ 091260254
General purpose enriched media with extensive applications for a vast range of mammalian cells including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes
TRUSTED_SUSTAINABILITY
Specifications
Product Type RPMI 1640 Medium
Sterility Sterile
Without Additives No Glutamine
Quantity 500 mL
pH 6.9 to 7.2
Content And Storage 15° to 30°C