Learn More
MP Biomedicals™ RPMI 1640 Medium
Shop All MP Biomedicals Products
Click to view available options
pH:
6.8 to 7.4
6.9 to 7.2
6.9 to 7.4
Content And Storage:
0° to 5°C
15° to 30°C
4°C
Unit Size:
100mL
500mL
Specifications
Specifications
| Product Type | RPMI 1640 Medium |
| Sterility | Sterile |
| Quantity | 100 mL |
| pH | 6.9 to 7.4 |
| Content And Storage | 4°C |