Learn More
Corning™ RPMI 1640, 1X
BIOPHARMA
Product Code. p-4591232
Click to view available options
Quantity:
6 x 500 mL
Product Type:
RPMI 1640, No L-glutamine, No Phenol Red
pH:
7 to 7.4
Unit Size:
Pack of 6
Product Code. 15363561
Brand: Corning™ 17105CV
Glutagro™ supplement provides greatly improved stability over the L-glutamine media supplement
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol. Cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.
  • Can be used for suspension cultures or monolayer human leukemic cells
  • This formulation is supplemented with glutaGRO™ in place of L-glutamine for improved stability
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 X
Product Type RPMI 1640, No L-glutamine, No Phenol Red
Without Additives No Glutamine, No Phenol Red
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 500 mL
Endotoxin Level <0.25EU/mL
Formulation L-Cystine/L-Methionine, Sodium Bicarbonate
pH 7 to 7.4
Content And Storage 2° to 8°C