missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RPLP0 (Human) Recombinant Protein
Human RPLP0 full-length ORF ( AAH15173, 1 a.a. - 317 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00006175-P03.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPREDRATWKSNYFLKIIQLLDDYPKCFIVGADNVGSKQMQQIRMSLRGKAVVLMGKNTMMRKAIRGHLENNPALEKLLPHIRGNVGFVFTKEDLTEIRDMLLANKVPAAARAGAIAPCEVTVPAQNTGLGPEKTSFFQALGITTKISRGTIEILSDVQLIKTGDKVGASEATLLNMLNISPFSFGLVIQQVFDNGSIYNPEVLDITEETLHSRFLEGVRNVASVCLQIGYPTVASVPHSIINGYKRVLALSVETDYTFPLAEKVKAFLADPSAFVAAAPVAAATTAAPAAAAAPAKVEAKEESEESDEDMGFGLFD
Specifications
AAH15173 | |
ribosomal protein, large, P0 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPLP0 | |
GST |
6175 | |
Wheat germ expression system | |
25 μg | |
L10E, MGC111226, MGC88175, P0, PRLP0, RPP0 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |