Learn More
RPL23A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16386-1-AP-20UL
 View more versions of this product
Product Code. 16845994
  • 1710.33 NOK / 20µL
Description
The function of this protein remains unknown.
Specifications
Specifications
RPL23A
Polyclonal
Unconjugated
RPL23A
60S ribosomal protein L23a, MDA20, ribosomal protein L23a, RPL23A
Rabbit
Antigen Affinity Chromatography
RUO
268449, 360572, 6147
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62750, P62751, P62752
Rpl23a
RPL23A Fusion Protein Ag9424
20 μL
Primary
Rat, Mouse, Human