missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPH3AL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15297-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene plays a direct regulatory role in calcium-ion-dependent exocytosis in both endocrine and exocrine cells and plays a key role in insulin secretion by pancreatic cells. This gene is likely a tumor suppressor. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
RPH3AL | |
Polyclonal | |
Unconjugated | |
RPH3AL | |
No C2 domains protein, NOC2, Rab effector Noc2, Rabphilin 3A like protein, RPH3AL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9501 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UNE2 | |
RPH3AL | |
RPH3AL Fusion Protein Ag7552 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |