missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24116-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
RNA polymerases transcribe nuclear genes for ribosomal RNA and are integral components of ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. RNA polymerase II (Pol II) synthesizes mRNA. RNA polymerase III (Pol III) transcribes class III genes, encoding a number of small ribosomal RNA molecules including tRNA and 5S rRNA. RPAP1 (RNA polymerase II associated protein 1) is a 1, 393 amino acid nuclear protein that is thought to connect Pol II to acetylated Histone H3 and regulators of protein complex formation. A member of the RPAP1 family, RPAP1 exists as three alternatively spliced isoforms that are encoded by a gene that maps to human chromosome 15q15.1.Specifications
RPAP1 | |
Polyclonal | |
Unconjugated | |
RPAP1 | |
KIAA1403, RPAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26015, 68925 | |
-20°C | |
Liquid |
Western Blot | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80TE0, Q9BWH6 | |
RPAP1 | |
RPAP1 Fusion Protein Ag7856 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |