missing translation for 'onlineSavingsMsg'
Learn More

RPA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16848194 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16848194 150 μL 150µL
16858194 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16848194

Brand: Proteintech 181441AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The single-stranded-DNA-binding proteins (SSBs) are essential for DNA function in prokaryotic and eukaryotic cells, mitochondria, phages and viruses. Replication protein A (RPA), a highly conserved eukaryotic protein, is a heterotrimeric SSB consisting of three subunits that play an important role in DNA replication, recombination and repair. RPA is one of the major damage- recognition structures involved in the early stage of nucleotide excision repair and may function in telomere maintece. The binding of human RPA (hRPA) to DNA involves molecular polarity, in which initial hRPA binding occurs on the 5′ side of a ssDNA substrate and then extends in the 3′ direction to create a stably bound hRPA. Widely expressed at low to intermediate levels,RPA 34 kDa subunit, which is also known as RPA4 (replication factor A protein 4), contains 261 amino acids, localizes to nucleus and is preferentially expressed in mucosa of colon and placenta.

Specifications

Antigen RPA4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPA4
Gene Accession No. Q13156
Gene Alias HSU24186, Replication factor A protein 4, replication protein A4, 34kDa, RF A protein 4, RP A p30, RPA4
Gene Symbols RPA4
Host Species Rabbit
Immunogen RPA4 Fusion Protein Ag12800
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29935
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.