Learn More
Emma Safety Footwear Roy Safety Shoes
Product Code. 15978821
Change view
Click to view available options
Size: 49
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15978821 Size: 49
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15978821 Supplier Emma Safety Footwear Supplier No. MM70054049
  • ONLINE EXCLUSIVE PRICE
    1160.25 NOK
    682.50 NOK / 1 pairs
    Save 477.75 NOK
    41% Off
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Versatile shoe with wipe clean uppers and an 'easy clean' sole. The shoe sculpts itself to the wearer's foot for comfort while an internal toe cap provides resilient protection against falling objects and unforeseen hazards.
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Roy
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 49
Toe Material Steel