Learn More
Thermo Scientific™ Round Test Tube Cells for Thermo Scientific Spectronic™ 20+ and 20D+ Spectrophotometers
Product Code. 11893774 Shop All Thermo Scientific Products
Change view
Click to view available options
Path Length:
25.4 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Path Length Unit Size
11893774 25.4 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11893774 Supplier Thermo Scientific™ Supplier No. 331782000
  • ONLINE EXCLUSIVE PRICE
    7145.00 NOK
    6634.90 NOK / Pack of 12
    Save 510.10 NOK
    7% Off
Estimated Shipment: 19-03-2027
Round Test Tube Cells for Thermo Scientific™ Spectronic™ 20+ and 20D+ Spectrophotometers
  • Optical class
  • Cat. No. 07-144-15 1/2 in. diameter test tube cell replacements for originals supplied with instrument Cat. No. 07-144-40 3/4 in. diameter test tube cells require 07-144-20 adapter and 07-144-24 light shield (sold separately)
  • Cat. No. 07-144-38 test tube cell is 1 in. diameter and requires 07-144-22 adapter and 07-144-24 light shield (sold separately)
  • Packaged in box of 12
TRUSTED_SUSTAINABILITY
Specifications
Type Test Tube
Material Optical Glass
Path Length 25.4 mm
Height (Metric) 150 mm
For Use With (Equipment) Thermo Scientific Spectronic 20 and 20D Spectrophotometers