Learn More
Cole-Parmer™ Round-Tapered Tip Tweezers
Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
12.7 cm
Unit Size:
Each
Description
- Finely crafted stainless steel tweezers
- Feature hand finished points for accuracy and excellent tip hardness
- Anti-glare finish reduces eye strain
- Anti-magnetic, anti-acid and acid resistant
Specifications
Specifications
| Tip Style | Round, Tapered |
| Length (English) | 5 in. |
| Material | Carbon Fiber |
| Length (Metric) | 12.7 cm |
| Product Type | Tweezers |
| Type | Soft and Strong Tweezers |
| Sterility | Sterile |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido