Learn More
Bochem™ Round Stainless Steel Wire Baskets with Handle
Specifications
Specifications
| Diameter (Metric) Outer | 120 mm |
| Weight (Metric) | 186 g |
| Includes | Handle |
| Product Type | Wire Basket |