Learn More
Epredia™ Round Specimen Clamps
Product Code. 12003069 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Clamp Case, 6 to 28 mm
Clamp Jaw Insert, 15 to 21mm
Clamp Jaw Insert, 22 to 28mm
Clamp Jaw Insert, 9 to 14.5mm
Unit Size:
Each
Product Code. Product Type unitSize
12003069 Clamp Jaw Insert, 15 to 21mm Each
12013069 Clamp Jaw Insert, 22 to 28mm Each
12023069 Clamp Jaw Insert, 9 to 14.5mm Each
12017219 Clamp Case, 6 to 28 mm Each
4 options
Product Code. 12003069
Brand: Epredia™ 546200
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure easy insertion and removal of round specimens with Epredia™ Round Specimen Clamps.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Clamp Jaw Insert, 15 to 21mm