Learn More
Gilac™ Round Selective Sorting Dustbin
Product Code. p-200009577
Click to view available options
Capacity (Metric):
50 L
75 L
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 17918851
Brand: Gilac™ G6132211
Only null left
Add to basket
Add to basket
Round Selective Sorting Dustbin is great as a highly durable recycling bin. Reinforced bottom with anti-wear skirt.
  • Round dustbin great as a recycling bin.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Capacity (Metric) 50 L
Diameter (Metric) Outer 610 mm
Diameter (English) Outer 24 in.
Capacity (English) 13.2 gal.
Color White
Height (Metric) 465 mm
Shape Round
Autoclavable Not Autoclavable
Height (English) 18.3 in.
Material HDPE
Product Type Dust Bin