Learn More
Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation
Product Code. 10396412
Change view
Click to view available options
Closure Type:
Not Included
Stopper
Unit Size:
Pack of 10
Product Code. Closure Type unitSize
10396412 Not Included Pack of 10
10767571 Stopper Pack of 10
2 options
Product Code. 10396412
Brand: Lenz Laborglasinstrumente™ 05700007
Only null left
Add to basket
Add to basket
Ideal for routine laboratory analysis and procedures. Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation are manufactured from durable DURAN™ borosilicate glass.
  • With round bottom and NS socket
  • Available without stopper, with PE stopper, or with hexagonal hollow glass stopper
  • Corrosion-free and resistant to thermal shock
Specifications
Diameter (Metric) 16 mm
Product Type Glass Tube
Material Borosilicate Glass
Height (Metric) 100 mm
Closure Type Not Included