Learn More
Korff™ Round Aluminum Foil Discs
Click to view available options
Diameter (Metric) Outer:
100 mm
120 mm
130 mm
150 mm
Unit Size:
Pack of 1000
Specifications
Specifications
| Diameter (Metric) Outer | 120 mm |
| Material | Aluminum |
| For Use With (Application) | Used for wide variety of laboratory tasks |