Learn More
Rotor/Stator Generators for IKA Ultra-Turrax™ Homogenizers
Product Code. 10499862 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
S 50 N-G 45 G
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10499862 S 50 N-G 45 G Each
1 options
Product Code. 10499862
Brand: IKA 0008003000
Specifications
Product Type Dispersing Element
Volume Range (Metric) 500 mL to 20 L
Diameter (Metric) 45 mm
Length (Metric) Shaft 300 mm
Material PTFE, 316L Stainless Steel
For Use With (Equipment) S 50 N-G 45 G
Model S 50N-G45-G
Packaging Quantity 1 Ea.
Sterilization Method Wet chemical