Learn More
Grant Instruments Rotor for MICROSPIN 12 PLUS
Click to view available options
Capacity:
16 x 0.2 mL Tubes
Unit Size:
Each
Description
Rotor for two 8 x 0.2 ml PCR strip rows or 16 individual 0.2 ml PCR tubes
Specifications
Specifications
| Material | Stainless Steel |
| Type | Fixed Angle Rotor |
| Capacity | 16 x 0.2 mL Tubes |
| Max. RCF | 15,588 x g |
| Max. Speed | 16,250 rpm |
| Max. Tube Length | 21 mm |
| Max. Tube Diameter | 7.5 mm |
| Net Weight | 100 g |
| For Use With (Equipment) | MICROSPIN 12 PLUS |
| Angle | 30° |
| Show More |