Learn More
OHAUS™ Rotor, Angle, 30 x 1.5/2.0 ml, ID, Sealable
Shop All Ohaus Products
Click to view available options
Capacity:
30 x 1.5/2.0 mL
Unit Size:
Each
Description
Requires
Frontier 5000 Series Multi Pro Centrifuge
- Capacity: 30 x 1.5/2.0 ml
- Max g-force: 20,595g
- Max speed: 14,000 rpm
Warranty
2 years
Specifications
Specifications
| Description | Rotor, Angle, 30x1.5/2.0ml, ID, Sealable |
| Material | Stainless Steel |
| Capacity | 30 x 1.5/2.0 mL |
| Max. RCF | 20,595 x g |
| Max. Speed | 14,000 rpm |
| Net Weight | 1.940 kg |
| For Use With (Equipment) | FC5714 |
| Angle | 45° |
| Includes | NA |