Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495, 30mL
Product Code. 12023874
Click to view available options
Capacity:
30 mL
Unit Size:
Each
Product Code. 12023874
Brand: Hettich™ 1365
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich carriers
  • For use with Rotofix and Universal centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Type Rotor Adapter
Tube Type Conical
Capacity 30 mL
Max. Tube Diameter 25 mm
Max. Tube Length 110 mm
For Use With (Equipment) Hettich Rotors with Carriers no. 1481, 1490 and 1495
Vessels per Adapter 1