Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Product Code. 12063874
Click to view available options
Capacity:
1 x 25 mL Tubes
12 mL
15 mL
27 x 1 mL Tubes
30 mL
4 x 4 to 10 mL
4 x 4 to 8.5 mL
5 x 1.6 to 10 mL Tubes
50 mL
7 x 1.1 to 1.4 mL
9 x 3 or 4 mL Tubes
Tube Type:
Blood/Urine Tubes
Conical
Rhesus Test Tubes
Round Bottom
Round Bottom, Conical, Blood/Urine Collection
Skirted Conical
Unit Size:
Each
Product Code. 12063874
Brand: Hettich™ 1457
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich carriers
  • For use with Rotofix and Universal centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Type Rotor Adapter
Tube Type Round Bottom, Conical, Blood/Urine Collection
Capacity 7 x 1.1 to 1.4 mL
Max. Tube Diameter 8 mm
Max. Tube Length 66 mm
For Use With (Equipment) Hettich Rotors with Carriers no. 1481, 1490 and 1495
Vessels per Adapter 7