Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Product Code. 10729663
Click to view available options
Capacity:
1 x 25 mL Tubes
12 mL
15 mL
27 x 1 mL Tubes
30 mL
4 x 4 to 10 mL
4 x 4 to 8.5 mL
5 x 1.6 to 10 mL Tubes
50 mL
7 x 1.1 to 1.4 mL
9 x 3 or 4 mL Tubes
Tube Type:
Blood/Urine Tubes
Conical
Rhesus Test Tubes
Round Bottom
Round Bottom, Conical, Blood/Urine Collection
Skirted Conical
Unit Size:
Each
Product Code. 10729663
Brand: Hettich™ 6311
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich carriers
  • For use with Rotofix and Universal centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Type Rotor Adapter
Tube Type Skirted Conical
Capacity 12 mL
Max. Tube Diameter 17 mm
Max. Tube Length 100 mm
For Use With (Equipment) Carriers no. 1481, 1490 and 1495
Vessels per Adapter 1
No. of Places 1