Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Product Code. 10695683
Change view
Click to view available options
Capacity:
1 x 25 mL Tubes
12 mL
15 mL
27 x 1 mL Tubes
30 mL
4 x 4 to 10 mL
4 x 4 to 8.5 mL
5 x 1.6 to 10 mL Tubes
50 mL
7 x 1.1 to 1.4 mL
9 x 3 or 4 mL Tubes
Tube Type:
Blood/Urine Tubes
Conical
Rhesus Test Tubes
Round Bottom
Round Bottom, Conical, Blood/Urine Collection
Skirted Conical
Unit Size:
Each
Product Code. Capacity Tube Type unitSize
10695683 50 mL Round Bottom Each
10658452 15 mL Conical Each
10457301 1 x 25 mL Tubes Round Bottom Each
12063864 27 x 1 mL Tubes Rhesus Test Tubes Each
12073864 9 x 3 or 4 mL Tubes Round Bottom Each
10173422 4 x 4 to 10 mL Round Bottom Each
10477111 5 x 1.6 to 10 mL Tubes Round Bottom, Conical, Blood/Urine Collection Each
12063874 7 x 1.1 to 1.4 mL Round Bottom, Conical, Blood/Urine Collection Each
10484432 4 x 4 to 8.5 mL Blood/Urine Tubes Each
10141053 30 mL Round Bottom Each
10075042 50 mL Skirted Conical Each
10729663 12 mL Skirted Conical Each
12 options
Product Code. 10695683
Brand: Hettich™ 4416
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich carriers
  • For use with Rotofix and Universal centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Type Rotor Adapter
Tube Type Round Bottom
Capacity 50 mL
Max. Tube Diameter 38 mm
Max. Tube Length 107 mm
For Use With (Equipment) Hettich Rotors with Carriers no. 1481, 1490 and 1495
Vessels per Adapter 1
No. of Places 1