Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Click to view available options
Capacity:
1 x 25 mL Tubes
12 mL
15 mL
27 x 1 mL Tubes
30 mL
4 x 4 to 10 mL
4 x 4 to 8.5 mL
5 x 1.6 to 10 mL Tubes
50 mL
7 x 1.1 to 1.4 mL
9 x 3 or 4 mL Tubes
Tube Type:
Blood/Urine Tubes
Conical
Rhesus Test Tubes
Round Bottom
Round Bottom, Conical, Blood/Urine Collection
Skirted Conical
Unit Size:
Each
Description
- Rotor adapter for Hettich carriers
- For use with Rotofix and Universal centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom |
| Capacity | 1 x 25 mL Tubes |
| Max. Tube Diameter | 24 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Hettich Rotors with Carriers no. 1481, 1490 and 1495 |
| Vessels per Adapter | 1 |