Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Description
- Rotor adapter for Hettich carriers
- For use with Rotofix and Universal centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom |
| Capacity | 30 mL |
| Max. Tube Diameter | 26 mm |
| Max. Tube Length | 95 mm |
| For Use With (Equipment) | Hettich Rotors with Carriers no. 1481, 1490 and 1495 |
| Vessels per Adapter | 1 |
| No. of Places | 1 |