Learn More
Hettich™ Rotor Adapter for Carriers no. 1398 and 1599
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1484
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10063842
  • 1125.00 NOK / Each
Estimated Shipment: 24-05-2024
Description
  • Rotor Adapter for Carriers no. 1398 and 1599
  • For use with ROTOFIX 32 A and Universal 320/320 R Centrifuges
Specifications
Specifications
Adapter
50 mL
115 mm
1
Conical
29 mm
Carriers no. 1398 and 1599 with ROTOFIX 32 A and Universal 320/320 R Centrifuges