Learn More
Hettich™ Rotor Adapter for Carriers
Click to view available options
For Use With (Equipment):
Carrier
Unit Size:
Each
Description
- Rotor Adapter
- Accessory for Hettich Carriers and Rotors
Specifications
Specifications
| Type | Rotor Adapter |
| For Use With (Equipment) | Carrier |
| Bore Size (Metric) | 17.6 mm |
| Vessels per Adapter | 42 Tubes |