Learn More
Hettich™ Rotor Adapter for Carrier no. 1752
Product Code. 11414988
Click to view available options
Capacity:
18 x 3 mL Tubes or 36 x 1.5 to 2 mL Tubes
3 x 45 to 75 mL Tubes
4 to 15 mL Tubes
Unit Size:
Each
Product Code. 11414988
Brand: Hettich™ 1765
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • For use with ROTINA 380/380 R
  • The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
  • Robust and steady high-capacity centrifuge
  • Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
  • Temperature control system with pre-cool function from -20 to +40°C
  • Versa-Rotor™ for multiple applications
  • All-steel construction
  • Quiet Operation
  • Easy-Lift™/Safety-Lock™ Lid
Specifications
Type Rotor Adapter
Tube Type Round Bottom
Capacity 3 x 45 to 75 mL Tubes
Max. Tube Diameter 35 mm
Max. Tube Length 105 mm
For Use With (Equipment) ROTINA 380/380 R Centrifuge with Carrier no. 1752
Vessels per Adapter 3