Learn More
Hettich™ Rotor Adapter for Carrier no. 1752
Click to view available options
Capacity:
18 x 3 mL Tubes or 36 x 1.5 to 2 mL Tubes
3 x 45 to 75 mL Tubes
4 to 15 mL Tubes
Unit Size:
Each
Description
- For use with ROTINA 380/380 R
- The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
- Robust and steady high-capacity centrifuge
- Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
- Temperature control system with pre-cool function from -20 to +40°C
- Versa-Rotor™ for multiple applications
- All-steel construction
- Quiet Operation
- Easy-Lift™/Safety-Lock™ Lid
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom, Microliter Tubes |
| Capacity | 18 x 3 mL Tubes or 36 x 1.5 to 2 mL Tubes |
| Max. Tube Diameter | 11 mm |
| Max. Tube Length | 60 mm |
| For Use With (Equipment) | ROTINA 380/380 R Centrifuge with Carrier no. 1752 |
| Vessels per Adapter | 18 |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido