Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Product Code. 10033942
Change view
Click to view available options
Capacity:
1 x 100 mL Tube
12 x 6 to 7 mL Tubes
14 x 3 mL
4 x 9 to 10 mL Tubes
50 mL
50 mL Tubes
7 x 1.6 to 7 mL Tubes
7 x 15 mL
Tube Type:
Conical
Conical Skirted
Round Bottom
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Tube Type unitSize
10033942 50 mL Tubes Conical Skirted Each
10392352 1 x 100 mL Tube Round Bottom Each
10318343 7 x 1.6 to 7 mL Tubes Conical Each
10659792 7 x 15 mL Round Bottom Each
12053874 14 x 3 mL Round Bottom Each
10272293 12 x 6 to 7 mL Tubes Round Bottom Each
10344443 4 x 9 to 10 mL Tubes Round Bottom Each
10347322 50 mL Conical Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10033942 Supplier Hettich™ Supplier No. 1737
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich Carriers
  • For use with Universal 320/320 R Centrifuge
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Type Adapter
Tube Type Conical Skirted
Capacity 50 mL Tubes
Max. Tube Diameter 29 mm
Max. Tube Length 115 mm
For Use With (Equipment) Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge
Vessels per Adapter 1
No. of Places 1