Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Product Code. 12053874
Change view
Click to view available options
Capacity:
1 x 100 mL Tube
12 x 6 to 7 mL Tubes
14 x 3 mL
4 x 9 to 10 mL Tubes
50 mL
50 mL Tubes
7 x 1.6 to 7 mL Tubes
7 x 15 mL
Tube Type:
Conical
Conical Skirted
Round Bottom
Unit Size:
Each
Product Code. Capacity Tube Type unitSize
12053874 14 x 3 mL Round Bottom Each
10392352 1 x 100 mL Tube Round Bottom Each
10318343 7 x 1.6 to 7 mL Tubes Conical Each
10659792 7 x 15 mL Round Bottom Each
10033942 50 mL Tubes Conical Skirted Each
10272293 12 x 6 to 7 mL Tubes Round Bottom Each
10344443 4 x 9 to 10 mL Tubes Round Bottom Each
10347322 50 mL Conical Each
8 options
Product Code. 12053874
Brand: Hettich™ 1433
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich Carriers
  • For use with Universal 320/320 R Centrifuge
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Type Adapter
Tube Type Round Bottom
Capacity 14 x 3 mL
Max. Tube Diameter 10 mm
Max. Tube Length 60 mm
For Use With (Equipment) Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge
Vessels per Adapter 14