Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Product Code. 10318343
Click to view available options
Capacity:
1 x 100 mL Tube
12 x 6 to 7 mL Tubes
14 x 3 mL
4 x 9 to 10 mL Tubes
50 mL
50 mL Tubes
7 x 1.6 to 7 mL Tubes
7 x 15 mL
Tube Type:
Conical
Conical Skirted
Round Bottom
Unit Size:
Each
Product Code. 10318343
Brand: Hettich™ 1438
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Rotor adapter for Hettich Carriers
  • For use with Universal 320/320 R Centrifuge
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Type Adapter
Tube Type Conical
Capacity 7 x 1.6 to 7 mL Tubes
Max. Tube Diameter 12 mm
Max. Tube Length 100 mm
For Use With (Equipment) Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge
Vessels per Adapter 7