Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Click to view available options
Capacity:
1 x 100 mL Tube
12 x 6 to 7 mL Tubes
14 x 3 mL
4 x 9 to 10 mL Tubes
50 mL
50 mL Tubes
7 x 1.6 to 7 mL Tubes
7 x 15 mL
Tube Type:
Conical
Conical Skirted
Round Bottom
Unit Size:
Each
Description
- Rotor adapter for Hettich Carriers
- For use with Universal 320/320 R Centrifuge
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Specifications
| Type | Adapter |
| Tube Type | Round Bottom |
| Capacity | 12 x 6 to 7 mL Tubes |
| Max. Tube Diameter | 12 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge |