Learn More
Hettich Lab Technology™ ROTINA 420 Centrifuge
Product Code. 10601494
Change view
Click to view available options
Capacity:
4 x 600 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10601494 4 x 600 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10601494 Supplier Hettich Lab Technology™ Supplier No. 4701
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
Well equipped for all centrifuging applications. ROTINA 420 Centrifuge is ideal for applications in clinical chemistry, in biotechnology and in life science research facilities.
  • ROTINA 420 Centrifuge
  • Compact benchtop centrifuge without rotor
  • Conceived for high sample throughput and high sample volumes
  • Maximum capacity of 4 x 600 mL
  • Holds 140 blood collection tubes or 16 microtitre plates
  • Compatible with a wide range of rotors and adapters
Specifications
Capacity 4 x 600 mL
Max. Speed 15,000 rpm
Max. RCF 24,400 x g
Refrigerated No
Height (English) Exterior 16.7 in.
Height (Metric) Exterior 423 mm
Width (English) Exterior 19.9 in.
Width (Metric) Exterior 506 mm
Certifications/Compliance EN / IEC 61326-1, Class B
Depth (English) 25.6 in.
Depth (Metric) 650 mm
Rotor Included No
Dimensions (HWD) 423 x 506 x 650 mm
Net Weight 75 kg
Run Time 1 sec. to 99 hr., 59 min., 59 sec., Continuous Run, Short Cycle Mode
Frequency 50/60 Hz
Voltage 200/240 V
Product Type Benchtop Centrifuge