Learn More
IKA™ ROTAVISC Vane Spindle
Product Code. 17277549 Shop All IKA Products
Change view
Click to view available options
Size:
12 x 83 x 12 mm (W x H x D)
14 x 106 x 14 mm (W x H) x D
149.5 x 35 x 35 mm (W x H x D)
22 x 124 x 22 mm (W x H x D)
8 x 97 x 8 mm (W x H x D)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17277549 149.5 x 35 x 35 mm (W x H x D) Each
17287549 22 x 124 x 22 mm (W x H x D) Each
17297549 14 x 106 x 14 mm (W x H) x D Each
17207559 8 x 97 x 8 mm (W x H x D) Each
17247569 12 x 83 x 12 mm (W x H x D) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17277549 Supplier IKA™ Supplier No. 0025006853
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Vane spindle designed for use with IKA™ ROTAVISC Viscometers.
  • Quick coupling
  • Hook hitch
  • M2.5 left-handed thread
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vane Spindle
For Use With (Equipment) IKA™ ROTAVISC Viscometer
Description VAN-SP-1 Vane Spindle
Material AISI 316L
Size 149.5 x 35 x 35 mm (W x H x D)