Learn More
IKA™ ROTAVISC Vane Spindle
Shop All IKA Products
Click to view available options
Size:
12 x 83 x 12 mm (W x H x D)
14 x 106 x 14 mm (W x H) x D
149.5 x 35 x 35 mm (W x H x D)
22 x 124 x 22 mm (W x H x D)
8 x 97 x 8 mm (W x H x D)
Unit Size:
Each
Description
- Quick coupling
- Hook hitch
- M2.5 left-handed thread
Specifications
Specifications
| Product Type | Vane Spindle |
| For Use With (Equipment) | IKA™ ROTAVISC Viscometer |
| Description | VAN-SP-5 Vane Spindle |
| Material | AISI 316L |
| Size | 12 x 83 x 12 mm (W x H x D) |