Learn More
Heidolph™ Condenser for Rotavac Vario Control
Product Code. 10121365 Shop All Heidolph Instruments Products
Change view
Click to view available options
Temperature (Metric) Max:
40°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Max Unit Size
10121365 40°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10121365 Supplier Heidolph™ Supplier No. 5910008400
  • ONLINE EXCLUSIVE PRICE
    7535.00 NOK
    7372.11 NOK / Pack of 1
    Save 162.89 NOK
    2% Off
Estimated Shipment: 13-11-2026
Condenser for Hei-VAC Vario Control, Vario Tec and Vario Station
Condenser for Hei-VAC Vario Control, Vario Tec and Vario Station
TRUSTED_SUSTAINABILITY
Specifications
Description Condenser
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Frequency 50/60 Hz
Enclosure Rating IP20
For Use With (Equipment) Rotavac Vario Control/Pumping Unit
Voltage 115 V