Learn More
Scientific Industries™ Rotator Genie Standard Platform with Bands
Product Code. 11391484 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Rotator Genie
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11391484 Rotator Genie Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11391484 Supplier Scientific Industries™ Supplier No. 83041458
  • ONLINE EXCLUSIVE PRICE
    2105.25 NOK
    1921.85 NOK / Pack of 1
    Save 183.40 NOK
    8% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Hold various types of vessels with this platform on the Rotator-Genie.
Standard platform with bands. Supplied with SI-210X. Can also be used on SI-220X.
TRUSTED_SUSTAINABILITY
Specifications
Model SI-2110
For Use With (Equipment) Rotator Genie