Learn More
Welch™ Rotary Vane Vacuum Pump
Product Code. 15341841 Shop All Welch Ilmvac Products
Change view
Click to view available options
Flow Rate:
0.9 CFM
5.6 CFM
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
15341841 0.9 CFM Each
15361841 5.6 CFM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15341841 Supplier Welch™ Supplier No. U1400N50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Welch Rotary Vane Vacuum Pump is ideal for pumping corrosive gases.
  • Corrosion-resistant parts are ideal for pumping corrosive gases such as Cl2, HBr, F3CCO2H, and HOAC
  • Pumps draw clean oil from top of oil reservoir, not from the bottom as do direct-drive pumps
  • Large oil reservoir dilutes contaminants
  • Lower pump speed (less than 580 rpm) results in less wear, cooler operation, and longer service life
  • Bubbler tube in the oil reservoir allows nitrogen gas purging, resulting in lower operating temperature, reduced corrosion rate, and extended pump oil life
TRUSTED_SUSTAINABILITY
Specifications
Horsepower 1/3 hp
Motor Type ODP
Amperage 3.5 amp
Includes Clamp, Centering Ring, NW Flange to Hose Barb Fitting, Pump Oil, and Instruction Manual
Dimensions (L x W x H) 45 x 22.9 x 32 cm (17.75 x 9 x 12.625 in.)
Model 1400N-50
Noise Level 59 dB
Weight (Metric) 26 kg
Temperature (English) Max. 145°F
Temperature (Metric) Max 63°C
Flow Rate 0.9 CFM
Frequency 50 Hz
Pump Type Rotary Vane Vacuum Pump
Oil Capacity 0.59 L
Voltage 230 V