Learn More
Welch Ilmvac™ Rotary Vane Pump Major Service Kit
Product Code. 10114957 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With:
CHEMVAC 8965
P 8 Z
PK 2 DC
Unit Size:
Each
Product Code. For Use With unitSize
10114957 CHEMVAC 8965 Each
10672933 PK 2 DC Each
10256937 P 8 Z Each
3 options
Product Code. 10114957
Brand: Welch Ilmvac™ 302079
Only null left
Specifications
For Use With CHEMVAC 8965
Includes Minor Kit Plus Vanes, Springs, Oil Glass
Quantity 1 Pack
Type Major Service Kit