Learn More
Fisherbrand™ Rotary Mixer Discs
Product Code. 12398448 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Rotary Mixer (RM-1 and RM-2)
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12398448 Rotary Mixer (RM-1 and RM-2) Each
1 options
Product Code. 12398448
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The range of accessories can also be autoclaved and allow for easy loading and tube removal.
  • Rotary Mixer Disc plain ready to accept Flask 'Pick n Mix' or Tube 'Pick n Mix' clips
  • Can be autoclaved and allow for easy loading and tube removal
  • The accessory is easy to fit onto the RM-1 locating on three pins then secured in place with thumb wheel
Specifications
For Use With (Equipment) Rotary Mixer (RM-1 and RM-2)