Learn More
Hirschmann Instruments™ rotarus™ Volume 100 Peristaltic Pump
Product Code. 15443583
Click to view available options
Motor Speed:
5 to 500 rpm
Unit Size:
Each
Product Code. 15443583
Brand: Hirschmann Instruments™ 9501562
Facilitates high flow rates conveniently and comprehensively in the laboratory. Hirschmann Instruments™ rotarus™ Volume 100 Peristaltic Pump offers continuous delivery in a dispensing pump across a broad range of applications.
  • Changeable pump heads, single and multichannel design
  • Rapid tool-free changing of pump heads Automatic pump head and hose detection with RFID technology Control panel located directly on pump housing TFT touchscreen with user-friendly menu
  • Calibratable
  • Method memory
  • Operation with 24 V DC USB and RS232 interface for PC link using interface (available as accessory)
  • Pump heads from other manufacturers can be fitted without difficulty using an adapter
  • Comprehensive accessory range
Specifications
Channels 1 to 12 (Depending on the pump head type used)
Motor Speed 5 to 500 rpm
Model volume 100
Motor Type DC
Dimensions (L x W x H) 270 x 100 x 220 mm
Weight (Metric) 10.1 kg
Voltage 100/240 VAC
Wattage 100 w
Frequency 50/60 Hz