Learn More
Hirschmann™ Rotarus™ MKF 60-16-2-4
Click to view available options
Capacity (Metric):
0.2 to 500 L/min
Unit Size:
Each
Description
- For regular stopper-less tubes with a WT of 1.6 mm
- 4 rollers and 2 channels
- Only an adapter plate is necessary for the connection between pump head and pump drive
- The pump head of the flow and volume pump drives is recognized by RFID technology
- The operation of the multi-channel head is made even safer by a finger protection
- Sturdy tubing cassettes with screws allow a fine adjustment of the flow rate
- Fixed distances between rollers and tubing bed allow the use with regular pump tubing without stoppers in a multi-channel pump head
- The pump head is made of durable PPS, a high-performance plastic with very high heat resistance, high chemical resistance and durability
Specifications
Specifications
| Accuracy | Approx. 5% |
| Capacity (Metric) | 0.2 to 500 L/min |
| Color | Black |
| Dimensions (L x W x H) | 88 x 115 x 98 mm |
| Flow Rate | Generally from 0.1 to 1350 mL/min. per channel |
| For Use With | Pump drive and tubing |
| Material | Polyphenylene Sulfide |
| No. of Rollers | 4 |
| Overall Height (Metric) | 98 mm |
| Overall Length (Metric) | 88 mm |
| Show More |