Learn More
Hettich™ ROTANTA 460 R Centrifuge
Product Code. 11425168
Change view
Click to view available options
Capacity:
4 x 750 mL
Unit Size:
Each
Product Code. Capacity unitSize
11425168 4 x 750 mL Each
1 options
Product Code. 11425168
Brand: Hettich™ 5660
Advanced laboratory technology makes a significant contribution to shortening processing times. ROTANTA Centrifuges are ideal for sample processing in clinics and laboratory centers.
  • Model ROTANTA 460 R
  • Refrigerated benchtop centrifuge
  • Enhanced performance and an extended range of accessories
  • fast, trouble-free operation in daily laboratory use
  • Maximum capacity of 3 liters per run
  • Without rotor
Specifications
Certifications/Compliance EN/IEC 61326-1, Class B
Capacity 4 x 750 mL
Max. RCF 244,000 x g
Max. Speed 15,000 rpm
Product Line ROTANTA 460 R
Refrigerated Yes
Run Time 1 sec. to 99 hr. 59 min. 59 sec.
Temperature Range -20°C to +40°C
Type Centrifuge
Dimensions (HWD) 456 x 554 x 706 mm
Net Weight 141 kg
Voltage 200/240 V
Frequency 50 Hz