Learn More
Hettich Lab Technology™ ROTANTA 460 Centrifuge
Click to view available options
Capacity:
4 x 750 mL
Unit Size:
Each
Description
- Enhanced performance and an extended range of accessories
- Fast, trouble-free operation in daily laboratory use
- Maximum capacity of 3 liters per run
Specifications
Specifications
| Certifications/Compliance | EN/IEC 61326-1, Class B |
| Capacity | 4 x 750 mL |
| Max. RCF | 244,000 x g |
| Max. Speed | 15,000 rpm |
| Product Line | ROTANTA 460 |
| Run Time | 1 sec. to 99 hr., 59 min., 59 sec. |
| Type | Centrifuge |
| Dimensions (HWD) | 456 x 554 x 706 mm |
| Width (English) Exterior | 21.8 in. |
| Width (Metric) Exterior | 554 mm |
| Show More |