Learn More
Rotaflo™ Stopcock Spare Parts
Product Code. 12967306
Change view
Click to view available options
Type:
Locking Ring
Stopcock Key
Stopcock Spare Parts
Plug Bore:
3 mm
6 mm
10 mm
Unit Size:
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Plug Bore Unit Size
12967306 Stopcock Key 6 mm Pack of 5
12947306 Locking Ring 3 mm Pack of 5
12852785 Locking Ring 6 mm Pack of 5
10213410 Stopcock Key 3 mm Pack of 5
12693022 Stopcock Key 10 mm Pack of 5
12643032 Stopcock Spare Parts 10 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12967306 Supplier Quickfit Supplier No. GP6K
  • ONLINE EXCLUSIVE PRICE
    2055.00 NOK
    1723.10 NOK / Pack of 5
    Save 331.90 NOK
    16% Off
Estimated Shipment: 11-01-2027
Replacement locking rings and keys for Quickfit Rotaflo™ stopcocks
Replacement keys and locking rings should be ordered separately (please see our website, or contact Customer Service)
Specifications
Type Stopcock Key
Plug Bore 6 mm
For Use With (Equipment) GP Rotaflo™ stopcock