Learn More
Rotaflo™ Stopcock Spare Parts
Product Code. 12693022
Change view
Click to view available options
Type:
Locking Ring
Stopcock Key
Stopcock Spare Parts
Plug Bore:
3 mm
6 mm
10 mm
Unit Size:
Pack of 5
Product Code. Type Plug Bore unitSize
12693022 Stopcock Key 10 mm Pack of 5
12947306 Locking Ring 3 mm Pack of 5
12852785 Locking Ring 6 mm Pack of 5
10213410 Stopcock Key 3 mm Pack of 5
12967306 Stopcock Key 6 mm Pack of 5
12643032 Stopcock Spare Parts 10 mm Pack of 5
6 options
Product Code. 12693022
Brand: Quickfit GP10K
Only null left
Add to basket
Add to basket
Replacement locking rings and keys for Quickfit Rotaflo™ stopcocks
Replacement keys and locking rings should be ordered separately (please see our website, or contact Customer Service)
TRUSTED_SUSTAINABILITY
Specifications
Type Stopcock Key
Plug Bore 10 mm
For Use With (Equipment) GP Rotaflo™ stopcock