Learn More
Roseoflavin, TRC
Specifications
Specifications
| Chemical Name or Material | Roseoflavin |
| CAS | 51093-55-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H24 N4 O6 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C19H24N4O6/c1-9-4-10-5-11-17(20-19(29)21-18(11)28)23(13(10)6-12(9)22(2)3)7-14(25)16(27)15(26)8-24/h4-6,14-16,24-27H,7-8H2,1-3H3,(H,21,28,29)/t14-,15+,16-/m0/s1 |
| SMILES | O[C@H]([C@H](O)[C@H](O)CO)CN(C1=NC2=O)C(C(N=C1C(N2)=O)=C3)=CC(N(C)C)=C3C |
| IUPAC Name | 8-(dimethylamino)-7-methyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]pyrimido[4,5-b]quinoline-2,4-dione |
| Molecular Weight (g/mol) | 404.42 |
| Formula Weight | 404.1696 |
| Show More |