Learn More
Eurofins DiscoverX Ros Protein, Active
Description
Protein tyrosine kinase ROS is a role for morphogenic induction and proliferation of epithelial cells in the kidney, intestine and lung. ROS may provide a signal transduction pathway for epithelial-mesenchymal interactions.
GenBank M34353. The recombinant protein contains the conflict V2262A with respect toGenBank M34353. This is reported inGenBank NM_002944 and Z98880.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 53.4 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥60% |
| Protein | Ros, active |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.